Ammonium 1-pyrrolidinecarbodithioate ≥95.0%

Supplier: TCI America

Synonyms: Pyrrolidinedithiocarbamate ammonium salt, 1-Pyrrolidinecarbodithioic acid, ammonium salt, Ammonium pyrrolidine-1-carbodithioate, Pyrrolidine-1-Dithiocarboxylic Acid Ammonium Salt, APCD

P0644-25G P0644-250G
TCP0644-025GEA 123.59 CAD
TCP0644-025G TCP0644-250G
Ammonium 1-pyrrolidinecarbodithioate ≥95.0%
Ammonium 1-pyrrolidinecarbodithioate

[Reagent for Atomic Absorption Analysis]
CAS Number: 5108-96-3
MDL Number: MFCD00012720
Molecular Formula: C5H9NS2
Molecular Weight: 164.29
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 144
Storage Temperature: <0°C


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Reagents for Nitric Oxide Research
Other Product Names / Synonyms: Ammonium Pyrrolidine-N-dithiocarbamate, 1-Pyrrolidinecarbodithioic Acid Ammonium Salt, Pyrrolidine-N-dithiocarbamic Acid Ammonium Salt, APDC

Formula: C₅H₁₂N₂S₂
MW: 146.3 · 18.0 g/mol
Melting Pt: 144 °C
MDL Number: MFCD00012720
CAS Number: 5108-96-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=678afd344c7ca5ee14b0f353166ff08d1bbf63ac&vid=b44558dcceffb3e162605437544ec2a3bd00d201&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR