Pyridoxine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

P0561-500G P0561-100G
TCP0561-500GEA 662.02 CAD
TCP0561-500G TCP0561-100G
Pyridoxine hydrochloride ≥98.0% (by HPLC, titration analysis)
Pyridoxine hydrochloride

CAS Number: 58-56-0
MDL Number: MFCD00012807
Molecular Formula: C8H11NO3
Molecular Weight: 205.64
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 207


Product Categories: Bioscience, Vitamins
Other Product Names / Synonyms: Adermin Hydrochloride, Vitamin B6 Hydrochloride

Formula: C₈H₁₂ClNO₃
Melting Pt: 207 °C
Storage Temperature: Ambient
MDL Number: MFCD00012807
CAS Number: 58-56-0
EINECS: 200-386-2
Merck Index: 13,08072
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6b24f859894fd64f84b4b44a3c66400bdb2f192&vid=a6b24f859894fd64f84b4b44a3c66400bdb2f192&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR