Benzyl propionate ≥99.0%

Supplier: TCI America

P0501-25ML P0501-500ML
TCP0501-025MLEA 34.76 CAD
TCP0501-025ML TCP0501-500ML
Benzyl propionate ≥99.0%
Benzyl propionate

CAS Number: 122-63-4
MDL Number: MFCD00027003
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 222
Flash Point (°C): 108
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: Propionic Acid Benzyl Ester

Formula: C₁₀H₁₂O₂
Boiling Pt: 222 °C
Density: 1.03 (20 °C)
Flash Pt: 108 °C
MDL Number: MFCD00027003
CAS Number: 122-63-4
EINECS: 204-559-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b55b34fe39b9041dbdd77b1515187407618bfbe3&vid=b55b34fe39b9041dbdd77b1515187407618bfbe3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR