Phthaloyl dichloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

P0405-100G
TCP0405-100GEA 95.61 CAD
TCP0405-100G
Phthaloyl dichloride ≥98.0% (by titrimetric analysis)
Phthaloyl dichloride

CAS Number: 88-95-9
MDL Number: MFCD00000666
Molecular Formula: C8H4Cl2O2
Molecular Weight: 203.02
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 276
Flash Point (°C): 151
Freezing point (°C): 12
Specific Gravity (20/20): 1.41


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Phthaloyl Dichloride

Formula: C₈H₄Cl₂O₂
Boiling Pt: 276 °C
Density: 1.41 (20 °C)
Flash Pt: 151 °C
MDL Number: MFCD00000666
CAS Number: 88-95-9
EINECS: 201-869-0
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8cd26e55e1856cde595d5076c16561ed3f9d5c58&vid=8cd26e55e1856cde595d5076c16561ed3f9d5c58&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR