o-Cresolphthalein complexone tetrasodium salt

Supplier: TCI America

Synonyms: o-Cresolphthalein Complexone sodium salt

P0344-1G
TCP0344-001GEA 221.2 CAD
TCP0344-001G
o-Cresolphthalein complexone tetrasodium salt
o-Cresolphthalein complexone tetrasodium salt

CAS Number: 62698-54-8
MDL Number: MFCD00066371
Form: Crystal
Color: Very Pale Yellow


Other Product Names / Synonyms: Cresolphthalein-bis(methyliminodiacetic Acid) Sodium Salt, o-Cresolphthalein Complexon Sodium Salt, Metal Phthalein Sodium Salt, Phthalein Purple Sodium Salt

Formula: C₃₂H₂₈N₂Na₄O₁₂
MW: 724.54 g/mol
MDL Number: MFCD00066371
CAS Number: 62698-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR