Pentacene, purified by sublimation

Supplier: TCI America

P0030-1G P0030-100MG
TCP0030-001GEA 470.84 CAD
TCP0030-001G TCP0030-100MG
Pentacene, purified by sublimation
Pentacene

CAS Number: 135-48-8
MDL Number: MFCD00003710
Molecular Formula: C22H14
Molecular Weight: 278.35
Form: Crystal
Color: Purple

Formula: C₂₂H₁₄
Melting Pt: 300 °C
Density: 1.3 g/cm³ (20 °C)
MDL Number: MFCD00003710
CAS Number: 135-48-8
EINECS: 205-193-7
Merck Index: 12,07240
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1fcca75eca7c6ca50aaf4678d80c520f67766afd&vid=ec5ca7a202e7d990135e840d8470a6c2d2e00190&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR