2,3,7,8,12,13,17,18-Octafluoro-5,10,15,20-tetrakis(pentafluorophenyl)porphyrin ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 2,3,7,8,12,13,17,18-Octafluoro-5,10,15,20-tetrakis(pentafluorophenyl)-21H,23H-porphine, Perfluoro-5,10,15,20-tetraphenyl-21H,23H-porphyrin

O0319-100MG
TCO0319-100MGEA 1495.94 CAD
TCO0319-100MG
2,3,7,8,12,13,17,18-Octafluoro-5,10,15,20-tetrakis(pentafluorophenyl)porphyrin ≥98.0% (by HPLC)
2,3,7,8,12,13,17,18-Octafluoro-5,10,15,20-tetrakis(pentafluorophenyl)porphyrin

CAS Number: 121399-88-0
Molecular Formula: C44H2F28N4
Molecular Weight: 1118.48
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Bioscience, Porphyrins
Other Product Names / Synonyms: Perfluoro-5,10,15,20-tetraphenyl-21H,23H-porphyrin, 2,3,7,8,12,13,17,18-Octafluoro-5,10,15,20-tetrakis(pentafluorophenyl)-21H,23H-porphine

Formula: C₄₄H₂F₂₈N₄
MW: 1118.48 g/mol
CAS Number: 121399-88-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64eba682f59e9ba39ec3d4b1cb1ce1e0101adc06&vid=64eba682f59e9ba39ec3d4b1cb1ce1e0101adc06&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR