3-n-Octylpyrrole ≥98.0%

Supplier: TCI America

Synonyms: 3-Octyl-1H-pyrrole

O0241-1G O0241-5G
TCO0241-001GEA 290.88 CAD
TCO0241-001G TCO0241-005G
3-n-Octylpyrrole ≥98.0%
3-n-Octylpyrrole

CAS Number: 118799-18-1
MDL Number: MFCD00191474
Molecular Formula: C12H21N
Molecular Weight: 179.31
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 111
Specific Gravity (20/20): 0.88

Formula: C₁₂H₂₁N
Boiling Pt: 111 °C
Density: 0.88 (20 °C)
MDL Number: MFCD00191474
CAS Number: 118799-18-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed63aa6281725fdc35d121cdd23e25788f051c72&vid=ed63aa6281725fdc35d121cdd23e25788f051c72&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR