Isoamyl n-octanoate (contains 2-Methylbutyl n-Octanoate)

Supplier: TCI America

Synonyms: Isopentyl n-Octanoate, n-Octanoic Acid Isoamyl Ester, Isopentyl octanoate

O0031-25ML
TCO0031-025MLEA 36.34 CAD
TCO0031-025ML
Isoamyl n-octanoate (contains 2-Methylbutyl n-Octanoate)
Isoamyl n-octanoate

CAS Number: 2035-99-6
MDL Number: MFCD00048917
Molecular Formula: C13H26O2
Molecular Weight: 214.35
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 250
Flash Point (°C): 85
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: Isopentyl n-Octanoate, n-Octanoic Acid Isoamyl Ester

Formula: C₁₃H₂₆O₂
MW: 214.35 g/mol
Boiling Pt: 250 °C
Density: 0.86 (20 °C)
Flash Pt: 85 °C
MDL Number: MFCD00048917
CAS Number: 2035-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR