2-Nitrobenzyl Cyclohexylcarbamate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: Cyclohexylcarbamic acid 2-Nitrobenzyl ester

N1052-1G
TCN1052-1GEA 560.27 CAD
TCN1052-1G
2-Nitrobenzyl Cyclohexylcarbamate ≥98.0% (by HPLC, total nitrogen)
2-Nitrobenzyl Cyclohexylcarbamate
Formula: C₁₄H₁₈N₂O₄
MW: 278.31 g/mol
MDL Number: MFCD22376660
CAS Number: 119137-03-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 102.0 to 106.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR