Dimethylammonium dichlorotri(mu-chloro)bis{(R)-(+)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}diruthenate(II)

Supplier: TCI America

N1019-200MG
TCN1019-1GEA 385.69 CAD
TCN1019-1G TCN1019-200MG
Dimethylammonium dichlorotri(mu-chloro)bis{(R)-(+)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis{(R)-(+)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}diruthenate(II)
Formula: C₁₀₆H₁₀₄Cl₅NP₄Ru₂
MDL Number: MFCD09753035
CAS Number: 944451-08-5

Order Now

Specification Test Results

Appearance Orange to Brown powder to crystal
IR to pass test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR