Noscapine hydrochloride hydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

N0963-5G
TCN0963-5GEA 172.22 CAD
TCN0963-5G
Noscapine hydrochloride hydrate ≥98.0% (by HPLC, titration analysis)
Noscapine hydrochloride hydrate
Formula: C₂₂H₂₃NO₇·HCl·xH₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00149223
CAS Number: 912-60-7
EINECS: 213-014-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +43.0 to +47.0 deg (C=0.5, 0.1mol/L HCl) (calcd.on anh.substance)
Water 2.0 to 6.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR