(+)-Nootkatone ≥97.0%

Supplier: TCI America

Synonyms: (4R,4aS,6R)-4,4a-Dimethyl-6-(prop-1-en-2-yl)-4,4a,5,6,7,8-hexahydronaphthalen-2(3H)-one

N0920-1G
TCN0920-1GEA 86.9 CAD
TCN0920-1G
(+)-Nootkatone ≥97.0%
(+)-Nootkatone

CAS Number: 4674-50-4
MDL Number: MFCD04974578
Molecular Formula: C15H22O
Molecular Weight: 218.34
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 36
Specific rotation [a]20/D: 173 deg (C=1, EtOH)
Storage Temperature: 0-10°C

Formula: C₁₅H₂₂O
Melting Pt: 36 °C
MDL Number: MFCD04974578
CAS Number: 4674-50-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a65cf36a9e3e482d2f0fedad552a9a791a62fb74&vid=a65cf36a9e3e482d2f0fedad552a9a791a62fb74&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR