New fuchsine for biochemical research

Supplier: TCI America

Synonyms: Basic Violet 2, New Fuchsin, 4-[(4-Amino-m-tolyl)(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-o-toluidine hydrochloride

N0873-5G
TCN0873-5GEA 66.04 CAD
TCN0873-5G
New fuchsine for biochemical research
New fuchsine

[for Biochemical Research]
CAS Number: 3248-91-7
MDL Number: MFCD00012567
Molecular Formula: C22H24ClN3
Molecular Weight: 365.91
Form: Crystal
Color: Deep Yellow Green


Other Product Names / Synonyms: Basic Violet 2

Formula: C₂₂H₂₄ClN₃
MW: 365.91 g/mol
MDL Number: MFCD00012567
CAS Number: 3248-91-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c596135365e2ac71763e0ecfdb89cf89e87c044c&vid=6fcfc086262bc7d3edc9972607a78219df7eaaa3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR