4-Nitro-D-phenylalanine hydrate ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

N0849-5G N0849-1G
TCN0849-5GEA 61.15 CAD
TCN0849-5G TCN0849-1G
4-Nitro-D-phenylalanine hydrate ≥97.0% (by HPLC, titration analysis)
4-Nitro-D-phenylalanine hydrate

CAS Number: 56613-61-7
MDL Number: MFCD00150962
Molecular Formula: C9H10N2O4
Molecular Weight: 210.19
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -6 deg (C=2, 1mol/L HCl)


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: H-D-Phe(4-NO2)-OH

Formula: C₉H₁₀N₂O₄·H₂O
MDL Number: MFCD00150962
CAS Number: 444777-67-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fd228643bb2b883ebf1b33179ad62d6f044d3ad&vid=6fd228643bb2b883ebf1b33179ad62d6f044d3ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR