4-Methoxy-3-nitrobenzaldehyde ≥98.0%

Supplier: TCI America

N0830-25G N0830-5G
TCN0830-25GEA 1140.44 CAD
TCN0830-25G TCN0830-5G
4-Methoxy-3-nitrobenzaldehyde ≥98.0%
4-Methoxy-3-nitrobenzaldehyde

CAS Number: 31680-08-7
MDL Number: MFCD00126498
Molecular Formula: C8H7NO4
Molecular Weight: 181.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 85


Other Product Names / Synonyms: 4-Methoxy-3-nitrobenzaldehyde

Formula: C₈H₇NO₄
MW: 181.15
Melting Pt: 85 °C
MDL Number: MFCD00126498
CAS Number: 31680-08-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3982f20576fe00e5cbece8d7222bfcf6c4f17af4&vid=3982f20576fe00e5cbece8d7222bfcf6c4f17af4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR