6-Nitroanthranilic acid ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

N0818-1G N0818-5G
TCN0818-1GEA 47.4 CAD
TCN0818-1G TCN0818-5G
6-Nitroanthranilic acid ≥97.0% (by HPLC, titration analysis)
6-Nitroanthranilic acid

CAS Number: 50573-74-5
MDL Number: MFCD00034785
Molecular Formula: C7H6N2O4
Molecular Weight: 182.14
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 173


Other Product Names / Synonyms: 2-Amino-6-nitrobenzoic Acid

Formula: C₇H₆N₂O₄
Melting Pt: 173 °C
MDL Number: MFCD00034785
CAS Number: 50573-74-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b18c96209ef5448aa70c4ce31fc5efadd6e5b4b9&vid=b18c96209ef5448aa70c4ce31fc5efadd6e5b4b9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR