cis-5-Norbornene exo-2,3-dicarboxylic anhydride ≥97.0% (by GC, titration analysis)

Supplier: TCI America

N0767-1G N0767-5G
TCN0767-1GEA 63.2 CAD
TCN0767-1G TCN0767-5G
cis-5-Norbornene exo-2,3-dicarboxylic anhydride ≥97.0% (by GC, titration analysis)
cis-5-Norbornene exo-2,3-dicarboxylic anhydride

CAS Number: 2746-19-2
Molecular Formula: C9H8O3
Molecular Weight: 164.16
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 144


Other Product Names / Synonyms: Bicyclo[2.2.1]hept-5-ene-exo-2,3-dicarboxylic Anhydride

Formula: C₉H₈O₃
Melting Pt: 144 °C
MDL Number: MFCD00630580
CAS Number: 2746-19-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fff73edb7fc2716934e4eb00240fc996096559f5&vid=fff73edb7fc2716934e4eb00240fc996096559f5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR