2,3-Norbornanedicarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

N0753-5G N0753-25G
TCN0753-5GEA 168.59 CAD
TCN0753-5G TCN0753-25G
2,3-Norbornanedicarboxylic acid ≥98.0% (by GC, titration analysis)
2,3-Norbornanedicarboxylic acid

CAS Number: 1724-08-9
Molecular Formula: C9H12O4
Molecular Weight: 184.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Bicyclo[2.2.1]heptane-2,3-dicarboxylic Acid

Formula: C₉H₁₂O₄
MDL Number: MFCD00520577
CAS Number: 1724-08-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff8b31f224609071e346b9ba2a5cbc37f891da81&vid=ff8b31f224609071e346b9ba2a5cbc37f891da81&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR