6-Nitropiperonal ≥97.0%

Supplier: TCI America

Synonyms: 6-Nitro-1,3-benzodioxole-5-carboxaldehyde

N0537-25G
TCN0537-025GEA 300.2 CAD
TCN0537-025G
6-Nitropiperonal ≥97.0%
6-Nitropiperonal

CAS Number: 712-97-0
MDL Number: MFCD00005829
Molecular Formula: C8H5NO5
Molecular Weight: 195.13
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 97


Other Product Names / Synonyms: 3,4-Methylenedioxy-6-nitrobenzaldehyde, 6-Nitro-1,3-benzodioxole-5-carboxaldehyde

Formula: C₈H₅NO₅
MW: 195.13 g/mol
Melting Pt: 97 °C
MDL Number: MFCD00005829
CAS Number: 712-97-0
EINECS: 211-926-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5da53db9303f314987396671d039e403db290744&vid=5da53db9303f314987396671d039e403db290744&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR