New fuchsine

Supplier: TCI America
Danger

N0478-25G
TCN0478-025GEA 70.74 CAD
TCN0478-025G
New fuchsine
New fuchsine

CAS Number: 3248-91-7
MDL Number: MFCD00012567
Molecular Formula: C22H24ClN3
Molecular Weight: 365.91
Form: Crystal
Color: Deep Yellow Green


Other Product Names / Synonyms: Basic Violet 2

Formula: C₂₂H₂₄ClN₃
Storage Temperature: Ambient
MDL Number: MFCD00012567
CAS Number: 3248-91-7
EINECS: 221-831-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR