2-Nitroethanol ≥93.0%

Supplier: TCI America
Warning

N0368-5G N0368-25G
TCN0368-005GEA 115.34 CAD
TCN0368-005G TCN0368-25G
2-Nitroethanol ≥93.0%
2-Nitroethanol

CAS Number: 625-48-9
MDL Number: MFCD00007405
Molecular Formula: C2H5NO3
Molecular Weight: 91.07
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Boiling point (°C): 197
Flash Point (°C): 113
Specific Gravity (20/20): 1.30

Formula: C₂H₅NO₃
Boiling Pt: 197 °C
Melting Pt: –80 °C
Density: 1.30 (20 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00007405
CAS Number: 625-48-9
EINECS: 210-895-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee09796a92078c3c675a054870c8cc69cf9553eb&vid=ee09796a92078c3c675a054870c8cc69cf9553eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR