Neutral red for synthesis

Supplier: TCI America

Synonyms: N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5

N0315-25G
TCN0315-025GEA 119.01 CAD
TCN0315-025G
Neutral red for synthesis
Neutral red

CAS Number: 553-24-2
MDL Number: MFCD00012651
Molecular Formula: C15H17ClN4
Molecular Weight: 288.78
Form: Crystal
Color: Deep Green
Melting point (°C): 290


Other Product Names / Synonyms: Neutral Red, Toluylene Red

Formula: C₁₅H₁₇ClN₄
MW: 288.78 g/mol
Melting Pt: 290
Storage Temperature: Ambient
MDL Number: MFCD00012651
CAS Number: 553-24-2
EINECS: 209-035-8
Merck Index: 12,06571
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ef65c7190d0d383e41d47eb9b8c3cd8dfb2f11a&vid=8ef65c7190d0d383e41d47eb9b8c3cd8dfb2f11a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR