5-Dimethylamino-2-nitrosophenol hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

N0261-5G
TCN0261-005GEA 353.76 CAD
TCN0261-005G TCN0261-025G
5-Dimethylamino-2-nitrosophenol hydrochloride ≥98.0% (by HPLC, titration analysis)
5-Dimethylamino-2-nitrosophenol hydrochloride

CAS Number: 41317-10-6
MDL Number: MFCD00060162
Molecular Formula: C8H10N2O2
Molecular Weight: 202.64
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Product Categories: Analytical Science, Bipyridyls, etc. (Chelating Agents)
Other Product Names / Synonyms: 2-Nitroso-5-dimethylaminophenol Hydrochloride

Formula: C₈H₁₀N₂O₂
MDL Number: MFCD00060162
CAS Number: 41317-10-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6ccde2dfad40424ba522972a036e98773a95fca&vid=b6ccde2dfad40424ba522972a036e98773a95fca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR