4'-Nitroacetophenone ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: p-Nitroacetophenone, 1-(4-Nitrophenyl)ethanone

N0111-25G N0111-500G N0111-100G
TCN0111-025GEA 51.43 CAD
TCN0111-025G TCN0111-500G TCN0111-100G
4'-Nitroacetophenone ≥98.0% (by GC)
4'-Nitroacetophenone

CAS Number: 100-19-6 MDL Number: MFCD00007355 Molecular Formula: C8H7NO3 Molecular Weight: 165.15 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 167 Melting point (°C): 79 Flash Point (°C): 149

Formula: C₈H₇NO₃
MW: 165.15 g/mol
Boiling Pt: 167 °C
Melting Pt: 79 °C
Flash Pt: 149
Storage Temperature: Ambient
MDL Number: MFCD00007355
CAS Number: 100-19-6
EINECS: 202-827-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f011bc6e5a711b6699e4d4e779efb6566a89760b&vid=f011bc6e5a711b6699e4d4e779efb6566a89760b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR