1,2-Naphthoquinone-4-sulfonic acid sodium salt ≥98.0% (by titrimetric analysis)

Supplier: TCI America

N0043-25G N0043-1G
TCN0043-025GEA 154.84 CAD
TCN0043-025G TCN0043-001G
1,2-Naphthoquinone-4-sulfonic acid sodium salt ≥98.0% (by titrimetric analysis)
1,2-Naphthoquinone-4-sulphonic acid sodium salt

CAS Number: 521-24-4
MDL Number: MFCD00001700
Molecular Formula: C10H6O5S
Molecular Weight: 260.19
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 1,2-Naphthoquinone-4-sulfonic Acid Sodium Salt

Formula: C₁₀H₅NaO₅S
MDL Number: MFCD00001700
CAS Number: 521-24-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7659c69281bea93dad840b96213013d2b08b8cd&vid=b7659c69281bea93dad840b96213013d2b08b8cd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR