Dipotassium-2-naphthol-6,8-disulfonate hydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

N0030-25G
TCN0030-025GEA 173.41 CAD
TCN0030-025G
Dipotassium-2-naphthol-6,8-disulfonate hydrate ≥98.0% (by HPLC, titration analysis)
Dipotassium-2-naphthol-6,8-disulfonate hydrate

CAS Number: 842-18-2
MDL Number: MFCD00066361
Molecular Formula: C10H8O7S2
Molecular Weight: 380.47
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: Dipotassium 7-Hydroxy-1,3-naphthalenedisulfonate, 2-Naphthol-6,8-disulfonic Acid Dipotassium Salt, G Acid Dipotassium Salt, 7-Hydroxy-1,3-naphthalenedisulfonic Acid Dipotassium Salt

Formula: C₁₀H₈O₇S₂
MDL Number: MFCD00066361
CAS Number: 842-18-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe374aca307e8e788d33ceebf037f3822263ba99&vid=fe374aca307e8e788d33ceebf037f3822263ba99&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR