4-(Methoxycarbonyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-(Methoxycarbonyl)benzeneboronic acid pinacol ester, Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

M3057-1G M3057-5G
TCM3057-1GEA 33.18 CAD
TCM3057-1G TCM3057-5G
4-(Methoxycarbonyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
4-(Methoxycarbonyl)phenylboronic acid pinacol ester
Formula: C₁₄H₁₉BO₄
MW: 262.11 g/mol
MDL Number: MFCD02179438
CAS Number: 171364-80-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 80.0 to 84.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR