2-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridylmethylthio]-1H-benzimidazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

M2800-1G M2800-5G
TCM2800-1GEA 97.96 CAD
TCM2800-1G TCM2800-5G
2-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridylmethylthio]-1H-benzimidazole ≥98.0% (by HPLC, titration analysis)
2-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridylmethylthio]-1H-benzimidazole
Formula: C₁₆H₁₄F₃N₃OS
Storage Temperature: Freezer
MDL Number: MFCD00834357
CAS Number: 103577-40-8

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 147.0 to 151.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR