Mangostine ≥97.0% (by HPLC)

Supplier: TCI America
Warning

M2793-100MG M2793-10MG
TCM2793-100MGEA 807.38 CAD
TCM2793-100MG TCM2793-10MG
Mangostine ≥97.0% (by HPLC)
Mangostine
Formula: C₂₄H₂₆O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00135200
CAS Number: 6147-11-1

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Melting point 179.0 to 183.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR