Methyl 4-Acetamido-2-methoxybenzoate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4-Acetamido-o-anisic acid methyl ester, 4-Acetamido-2-methoxybenzoic acid methyl ester, Methyl 4-Acetamido-o-anisate, Methyl 4-acetamido-2-methoxybenzoate

M2776-25G M2776-5G
TCM2776-25GEA 137.46 CAD
TCM2776-25G TCM2776-5G
Methyl 4-Acetamido-2-methoxybenzoate ≥98.0% (by GC)
Methyl 4-Acetamido-2-methoxybenzoate
Formula: C₁₁H₁₃NO₄
MW: 223.23 g/mol
MDL Number: MFCD00065258
CAS Number: 4093-29-2
EINECS: 223-839-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 128.0 to 132.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR