Methyl-2,5-dichloronicotinate ≥98.0% (by GC)

Supplier: TCI America
Warning

M2752-1G M2752-200MG
TCM2752-1GEA 158.63 CAD
TCM2752-1G TCM2752-200MG
Methyl-2,5-dichloronicotinate ≥98.0% (by GC)
Methyl 2,5-dichloronicotinate
Formula: C₇H₅Cl₂NO₂
MDL Number: MFCD03844857
CAS Number: 67754-03-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 42.0 to 46.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR