6-Methoxyquinoline-4-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: Quininic acid

M2751-1G
TCM2751-1GEA 375.25 CAD
TCM2751-1G
6-Methoxyquinoline-4-carboxylic acid ≥98.0% (by HPLC, titration analysis)
6-Methoxyquinoline-4-carboxylic acid
Formula: C₁₁H₉NO₃
MW: 203.20 g/mol
Melting Pt: 280°C
MDL Number: MFCD00024013
CAS Number: 86-68-0
EINECS: 237-014-3
Merck Index: 14,08062

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR