Methyl-3-iodo-4-methylbenzoate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Methyl 3-iodo-4-methylbenzoate

M2694-25G M2694-5G
TCM2694-25GEA 201.61 CAD
TCM2694-25G TCM2694-5G
Methyl-3-iodo-4-methylbenzoate ≥98.0% (by GC)
Methyl 3-iodo-4-methylbenzoate
Formula: C₉H₉IO₂
MW: 276.07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00230583
CAS Number: 90347-66-3

Order Now

Specification Test Results

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR