Monomethyl-trans-1,4-cyclohexanedicarboxylate ≥97.0% (by GC, titration analysis)

Supplier: TCI America

M2526-25G M2526-5G
TCM2526-25GEA 278.08 CAD
TCM2526-25G TCM2526-5G
Monomethyl-trans-1,4-cyclohexanedicarboxylate ≥97.0% (by GC, titration analysis)
Monomethyl-trans-1,4-cyclohexanedicarboxylate

CAS Number: 15177-67-0
MDL Number: MFCD01311247
Molecular Formula: C9H14O4
Molecular Weight: 186.21
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 128


Other Product Names / Synonyms: trans-1,4-Cyclohexanedicarboxylic Acid Monomethyl Ester, Methyl Hydrogen trans-1,4-Cyclohexanedicarboxylate

Formula: C₉H₁₄O₄
Melting Pt: 128 °C
MDL Number: MFCD01311247
CAS Number: 15177-67-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3428527ecc3c698888bb8988736dc53e92ec10ef&vid=3428527ecc3c698888bb8988736dc53e92ec10ef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR