Methyl-3-methyl-2-furoate ≥98.0% (by GC)

Supplier: TCI America

M2377-1G
TCM2377-1GEA 72.68 CAD
TCM2377-1G
Methyl-3-methyl-2-furoate ≥98.0% (by GC)
Methyl 3-methyl-2-furoate

CAS Number: 6141-57-7
MDL Number: MFCD00014110
Molecular Formula: C7H8O3
Molecular Weight: 140.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 74
Melting point (°C): 38
Flash Point (°C): 82


Other Product Names / Synonyms: 3-Methyl-2-furancarboxylic Acid Methyl Ester, Methyl 3-Methyl-2-furoate, 3-Methyl-2-furoic Acid Methyl Ester

Formula: C₇H₈O₃
Boiling Pt: 74 °C
Melting Pt: 38 °C
Flash Pt: 82 °C
MDL Number: MFCD00014110
CAS Number: 6141-57-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d723871997b53773f008982062d3f204c35fc9b&vid=5d723871997b53773f008982062d3f204c35fc9b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR