Myoseverin ≥98.0% (by HPLC)

Supplier: TCI America

M2373-10MG
TCM2373-10MGEA 282.66 CAD
TCM2373-10MG
Myoseverin ≥98.0% (by HPLC)
Myoseverin

CAS Number: 267402-71-1
MDL Number: MFCD02683596
Molecular Formula: C24H28N6O2
Molecular Weight: 432.53
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 108
Storage Temperature: <0°C


Product Categories: Bioscience, Regenerative Medicine Research Reagents
Other Product Names / Synonyms: 9-Isopropyl-2,6-bis(4-methoxybenzylamino)purine

Formula: C₂₄H₂₈N₆O₂
Melting Pt: 108 °C
MDL Number: MFCD02683596
CAS Number: 267402-71-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab503663df2453eb0df8b8d99756a09d69f36ff2&vid=ab503663df2453eb0df8b8d99756a09d69f36ff2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR