2-Methoxyethyl methacrylate ≥98.0% (by GC) stabilized

Supplier: TCI America

M2282-25G M2282-500G
TCM2282-25GEA 79 CAD
TCM2282-25G TCM2282-500G
2-Methoxyethyl methacrylate ≥98.0% (by GC) stabilized
2-Methoxyethyl methacrylate

(stabilized with MEHQ)
CAS Number: 6976-93-8
MDL Number: MFCD00048122
Molecular Formula: C7H12O3
Molecular Weight: 144.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 65
Flash Point (°C): 66
Specific Gravity (20/20): 0.99


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Ethylene Glycol Monomethyl Ether Methacrylate, Methacrylic Acid 2-Methoxyethyl Ester

Formula: C₇H₁₂O₃
Boiling Pt: 65 °C
Density: 0.99 (20 °C)
Flash Pt: 66 °C
MDL Number: MFCD00048122
CAS Number: 6976-93-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6db032a7a5ad5643fd023d4284960aa755a9b38&vid=f6db032a7a5ad5643fd023d4284960aa755a9b38&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR