Methyl trans-p-coumarate ≥98.0% (by GC)

Supplier: TCI America

M2259-25G M2259-5G
TCM2259-25GEA 360.24 CAD
TCM2259-25G TCM2259-5G
Methyl trans-p-coumarate ≥98.0% (by GC)
Methyl trans-p-coumarate

CAS Number: 19367-38-5
MDL Number: MFCD00157167
Molecular Formula: C10H10O3
Molecular Weight: 178.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 139
Specific rotation [a]20/D: 12.1 deg (C=0.12, MeOH)
Lambda max.: 315 nm (MeOH)


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: trans-p-Coumaric Acid Methyl Ester, trans-4-Hydroxycinnamic Acid Methyl Ester, Methyl trans-4-Hydroxycinnamate

Formula: C₁₀H₁₀O₃
Melting Pt: 139 °C
MDL Number: MFCD00157167
CAS Number: 19367-38-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de9115e2e599a67bd84abf937886de30c38e78c2&vid=de9115e2e599a67bd84abf937886de30c38e78c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR