Methylmagnesium bromide ∼30% ∼3 mol/L in MeTHF (2-methyltetrahydrofuran)

Supplier: TCI America
Danger

M2237-250G
TCM2237-250GEA 255.96 CAD
TCM2237-250G
Methylmagnesium bromide ∼30% ∼3 mol/L in MeTHF (2-methyltetrahydrofuran)
Methylmagnesium bromide
Formula: CH₃BrMg
MW: 119.24 g/mol
MDL Number: MFCD00000041
CAS Number: 75-16-1
EINECS: 200-844-1
UN: 3399
ADR: 4.3 I

Order Now

Specification Test Results

Concentration (sec Butanol method) 30.0 to 34.5 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR