Methyl-3-methyl-2-nitrobenzoate ≥98.0% (by GC)

Supplier: TCI America

M2213-25G M2213-5G
TCM2213-25GEA 186.76 CAD
TCM2213-25G TCM2213-5G
Methyl-3-methyl-2-nitrobenzoate ≥98.0% (by GC)
Methyl 3-methyl-2-nitrobenzoate

CAS Number: 5471-82-9
MDL Number: MFCD01003947
Molecular Formula: C9H9NO4
Molecular Weight: 195.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 74


Other Product Names / Synonyms: 3-Methyl-2-nitrobenzoic Acid Methyl Ester

Formula: C₉H₉NO₄
Melting Pt: 74 °C
MDL Number: MFCD01003947
CAS Number: 5471-82-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1908fd08b0b4252f9ca57cbd8f580cf5b5c19645&vid=1908fd08b0b4252f9ca57cbd8f580cf5b5c19645&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR