1-Methyl-1H-indazole-3-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M2143-5G
TCM2143-1GEA 188.78 CAD
TCM2143-1G TCM2143-5G
1-Methyl-1H-indazole-3-carboxylic acid ≥98.0% (by GC, titration analysis)
1-Methyl-1H-indazole-3-carboxylic acid

CAS Number: 50890-83-0
MDL Number: MFCD00272569
Molecular Formula: C9H8N2O2
Molecular Weight: 176.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 216

Formula: C₉H₈N₂O₂
MW: 176.17 g/mol
Melting Pt: 216 °C
MDL Number: MFCD00272569
CAS Number: 50890-83-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=879798b52bc1d234385f785e356735b8eaeb01ad&vid=879798b52bc1d234385f785e356735b8eaeb01ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR