1-Methylbenzotriazole ≥98.0% (by GC)

Supplier: TCI America

M2121-5G
TCM2121-5GEA 113.76 CAD
TCM2121-5G
1-Methylbenzotriazole ≥98.0% (by GC)
1-Methylbenzotriazole

CAS Number: 13351-73-0
MDL Number: MFCD00014572
Molecular Formula: C7H7N3
Molecular Weight: 133.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 155
Melting point (°C): 64
Lambda max.: 285 nm (Cyclohexane)

Formula: C₇H₇N₃
Boiling Pt: 155 °C
Melting Pt: 64 °C
MDL Number: MFCD00014572
CAS Number: 13351-73-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=370023a97d75a4e499461578f9a74d4ebe521356&vid=370023a97d75a4e499461578f9a74d4ebe521356&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR