4-Nitro-2-picoline-N-oxide ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M2114-25G M2114-5G
TCM2114-25GEA 361.82 CAD
TCM2114-25G TCM2114-5G
4-Nitro-2-picoline-N-oxide ≥98.0% (by GC, titration analysis)
4-Nitro-2-picoline N-oxide

CAS Number: 5470-66-6
MDL Number: MFCD00014625
Molecular Formula: C6H6N2O3
Molecular Weight: 154.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 156
Lambda max.: 333 nm (EtOH)


Other Product Names / Synonyms: 4-Nitro-2-picoline N-Oxide

Formula: C₆H₆N₂O₃
Melting Pt: 156 °C
MDL Number: MFCD00014625
CAS Number: 5470-66-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6906c928478c023b8c087ca1f146c1f8faeac62f&vid=6906c928478c023b8c087ca1f146c1f8faeac62f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR