Methyl-β-D-ribofuranoside ≥97.0%

Supplier: TCI America

M1965-1G M1965-5G
TCM1965-1GEA 75.84 CAD
TCM1965-1G TCM1965-5G
Methyl-β-D-ribofuranoside ≥97.0%
Methyl-beta-D-ribofuranoside

CAS Number: 7473-45-2
MDL Number: MFCD00047075
Molecular Formula: C6H12O5
Molecular Weight: 164.16
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 78
Specific rotation [a]20/D: -51 deg (C=2, H2O)

Formula: C₆H₁₂O₅
Melting Pt: 78 °C
MDL Number: MFCD00047075
CAS Number: 7473-45-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fceed3318f164fbeb1bda0989377696c138663bd&vid=fceed3318f164fbeb1bda0989377696c138663bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR