Methyl Indole-2-carboxylate ≥98.0%

Supplier: TCI America

M1749-5G M1749-25G
TCM1749-5GEA 52.14 CAD
TCM1749-5G TCM1749-25G
Methyl Indole-2-carboxylate ≥98.0%
Methyl Indole-2-carboxylate

CAS Number: 1202-04-6
MDL Number: MFCD00460779
Molecular Formula: C10H9NO2
Molecular Weight: 175.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 153


Other Product Names / Synonyms: Indole-2-carboxylic Acid Methyl Ester

Formula: C₁₀H₉NO₂
Melting Pt: 153 °C
MDL Number: MFCD00460779
CAS Number: 1202-04-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f59507100298ad471d7de370bdd831b0c9f5ef7d&vid=f59507100298ad471d7de370bdd831b0c9f5ef7d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR