2-Methyl-4,5-diphenylthiazole 97,0

Supplier: TCI America

M1705-5G
TCM1705-5GEA 173.48 CAD
TCM1705-5G
2-Methyl-4,5-diphenylthiazole 97,0
2-Methyl-4,5-diphenylthiazole

CAS Number: 3755-83-7
Molecular Formula: C16H13NS
Molecular Weight: 251.35
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 52

Formula: C₁₆H₁₃NS
MW: 251.35 g/mol
Melting Pt: 52 °C
MDL Number: MFCD00186396
CAS Number: 3755-83-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9b2b04f0d2b70826aeb2aef39442dcee9c56592&vid=f9b2b04f0d2b70826aeb2aef39442dcee9c56592&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR