4'-Methoxyformanilide ≥98.0%

Supplier: TCI America

Synonyms: N-(4-Methoxyphenyl)formamide, N-Formyl-4-methoxyaniline

M1623-5G
TCM1623-5GEA 249.64 CAD
TCM1623-5G
4'-Methoxyformanilide ≥98.0%
4'-Methoxyformanilide

CAS Number: 5470-34-8
MDL Number: MFCD00086774
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 159
Melting point (°C): 80


Other Product Names / Synonyms: N-Formyl-4-methoxyaniline, N-(4-Methoxyphenyl)formamide

Formula: C₈H₉NO₂
MW: 151.17 g/mol
Boiling Pt: 159 °C
Melting Pt: 80 °C
MDL Number: MFCD00086774
CAS Number: 5470-34-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c3ddf7865f8598bb1af57ba56dd2f21e659f46f&vid=0c3ddf7865f8598bb1af57ba56dd2f21e659f46f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR