Methyl-4-(1,1-dioxothiomorpholino)benzoate ≥98.0%

Supplier: TCI America

M1587-1G
TCM1587-1GEA 875.32 CAD
TCM1587-1G
Methyl-4-(1,1-dioxothiomorpholino)benzoate ≥98.0%
Methyl-4-(1,1-dioxothiomorpholino)benzoate

CAS Number: 451485-76-0
Molecular Formula: C12H15NO4S
Molecular Weight: 269.32
Purity/Analysis Method: >98.0% (GC)
Form: Crystal


Other Product Names / Synonyms: 4-(1,1-Dioxothiomorpholino)benzoic Acid Methyl Ester, 4-[4-(Methoxycarbonyl)phenyl]thiomorpholine 1,1-Dioxide

Formula: C₁₂H₁₅NO₄S
CAS Number: 451485-76-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac94353b26d92e441acb3846a024efc83dfe50ff&vid=ac94353b26d92e441acb3846a024efc83dfe50ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR