(3-Mercaptopropyl)triethoxysilane ≥96.0%

Supplier: TCI America

Synonyms: 3-(Triethoxysilyl)propanethiol, 3-Mercaptopropyltriethoxysilane

M1505-25G M1505-500G M1505-100G
TCM1505-25GEA 56.25 CAD
TCM1505-25G TCM1505-500G TCM1505-100G
(3-Mercaptopropyl)triethoxysilane ≥96.0%
(3-Mercaptopropyl)triethoxysilane

CAS Number: 14814-09-6
MDL Number: MFCD00053924
Molecular Formula: C9H22O3SSi
Molecular Weight: 238.42
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 115
Flash Point (°C): 88
Specific Gravity (20/20): 0.99


Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: 3-(Triethoxysilyl)propanethiol

Formula: HSH₂CCH₂CH₂Si(OCH₂CH₃)₃
Boiling Pt: 115 °C
Density: 0.99 (20 °C)
Flash Pt: 88 °C
MDL Number: MFCD00053924
CAS Number: 14814-09-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11fa23e48013f81b4cbf283b4f4bd0cf07d75b92&vid=11fa23e48013f81b4cbf283b4f4bd0cf07d75b92&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR